| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05004585.hg.1 |
TC05000336.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05004585.hg.1 |
ENST00000296755.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05004585.hg.1 |
ENST00000504183.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05004585.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05004584.hg.1 |
TC05000336.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05004584.hg.1 |
ENST00000296755.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05004584.hg.1 |
ENST00000504183.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05004584.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05004582.hg.1 |
TC05000336.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05004582.hg.1 |
ENST00000296755.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05004582.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05004586.hg.1 |
TC05000336.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05004586.hg.1 |
ENST00000296755.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05004586.hg.1 |
ENST00000504183.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05004586.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000504183.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000504183.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000504183.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000504183.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000504492.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000504492.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000504492.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000504492.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000504492.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000511641.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000511641.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000511641.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000511641.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000511641.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000511641.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000511641.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000512974.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000512974.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000512974.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000512974.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000512974.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000512974.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000513526.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000513526.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000513526.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000513526.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000513526.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000513526.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000513526.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
ENST00000513526.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007738.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05004581.hg.1 |
TC05000336.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05004581.hg.1 |
ENST00000296755.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05004581.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500152704.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152704.hg.1 |
ENST00000504183.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152704.hg.1 |
ENST00000511641.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152704.hg.1 |
ENST00000512974.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152704.hg.1 |
ENST00000513526.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152704.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500152704.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152704.hg.1 |
ENST00000504183.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152704.hg.1 |
ENST00000511641.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152704.hg.1 |
ENST00000512974.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152704.hg.1 |
ENST00000513526.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152704.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500152704.hg.1 |
TC0500007738.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000504183.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000504183.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000504183.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000504183.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000504492.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000504492.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000504492.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000504492.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000504492.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000511641.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000511641.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000511641.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000511641.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000511641.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000511641.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000511641.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000512974.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000512974.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000512974.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000512974.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000512974.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000512974.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000513526.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000513526.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000513526.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000513526.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000513526.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000513526.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000513526.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
ENST00000513526.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007738.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500152700.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152700.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500152700.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152700.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500152700.hg.1 |
TC0500007738.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500152701.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152701.hg.1 |
ENST00000512974.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152701.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500152701.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152701.hg.1 |
ENST00000512974.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152701.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500152701.hg.1 |
TC0500007738.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500152703.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152703.hg.1 |
ENST00000504183.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152703.hg.1 |
ENST00000511641.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152703.hg.1 |
ENST00000512974.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152703.hg.1 |
ENST00000513526.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152703.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500152703.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152703.hg.1 |
ENST00000504183.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152703.hg.1 |
ENST00000511641.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152703.hg.1 |
ENST00000512974.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152703.hg.1 |
ENST00000513526.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152703.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500152703.hg.1 |
TC0500007738.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500152705.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152705.hg.1 |
ENST00000504183.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152705.hg.1 |
ENST00000511641.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152705.hg.1 |
ENST00000512974.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152705.hg.1 |
ENST00000513526.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152705.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500152705.hg.1 |
ENST00000296755.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152705.hg.1 |
ENST00000504183.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152705.hg.1 |
ENST00000511641.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152705.hg.1 |
ENST00000512974.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152705.hg.1 |
ENST00000513526.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152705.hg.1 |
NM_005909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500152705.hg.1 |
TC0500007738.hg.1 |
Clariom D TC |