Assay Details
| Gene Symbol: ANKRA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023039.4 | NP_075526.1 | 4 - 5 | 1168 | 99 | |
| XM_005248560.2 | XP_005248617.1 | 4 - 5 | 1200 | 99 | ||
| XM_017009678.1 | XP_016865167.1 | 4 - 5 | 1026 | 99 | ||
| GenBank mRNA | AF251051.1 | - | 4 - 5 | 1156 | 99 | |
| AF314032.1 | - | 3 - 4 | 588 | 99 | ||
| AK022876.1 | - | 4 - 5 | 915 | 99 | ||
| BC012917.1 | - | 4 - 5 | 1034 | 4452497 | 99 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001472.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017009678.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000504641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000504641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000504641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000504641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000504641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000504641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000504641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000504641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000505301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000505301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000505301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000505301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000505301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000505301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000506130.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000506130.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000506130.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000506130.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000509433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000509433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000509433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000509433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000509433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000514916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000514916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000514916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000514916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000515249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000515249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000515249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000515249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000515249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000515249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000515249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000515804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000515804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000515804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000515804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000515804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000515804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000515804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000515804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000515804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | ENST00000515804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05020275.hg.1 | TC05001472.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05020275.hg.1 | ENST00000296785.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05020275.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000504641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000504641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000504641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000504641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000504641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000504641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000504641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000504641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000505301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000505301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000505301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000505301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000505301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000505301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000506130.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000506130.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000506130.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000506130.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000509433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000509433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000509433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000509433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000509433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000514916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000514916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000514916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000514916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000515249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000515249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000515249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000515249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000515249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000515249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000515249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000515804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000515804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000515804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000515804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000515804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000515804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000515804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000515804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000515804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | ENST00000515804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011125.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500177235.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500177235.hg.1 | ENST00000504641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500177235.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500177235.hg.1 | ENST00000296785.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500177235.hg.1 | ENST00000504641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500177235.hg.1 | NM_023039.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500177235.hg.1 | TC0500011125.hg.1 | Clariom D TC |

ANKRA2
Genomic Map