| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000311364.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000311364.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000311364.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000311364.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000311364.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000311364.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000311364.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000311364.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000311364.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000311364.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000311364.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000458310.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000458310.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000458310.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000458310.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000458310.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000458310.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000458310.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000458310.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000458310.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000458310.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000458310.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000504893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000504893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000504893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000504893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000504893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000506212.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000506212.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000506212.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000506212.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000511684.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000511684.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000511684.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000511684.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000511684.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000511684.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000513950.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000513950.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
ENST00000513950.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_001131065.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_001131065.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_001131065.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_001131065.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_001131065.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_001131065.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_001131065.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_001131065.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_001131065.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_001131065.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_001131065.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_001131066.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_001131066.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_001131066.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_001131066.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_001131066.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_001131066.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_001131066.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_001131066.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_001131066.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_001131066.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_173362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_173362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_173362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_173362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_173362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_173362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_173362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_173362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_173362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013200.hg.1 |
NM_173362.3 |
Refseq |