Assay Details
| Gene Symbol: CAMK1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153498.3 | NP_705718.1 | 11 | 3914 | 146 | |
| XM_006717481.3 | XP_006717544.1 | 11 | 4178 | 146 | ||
| XM_006717482.3 | XP_006717545.1 | 11 | 3900 | 146 | ||
| XM_006717483.3 | XP_006717546.1 | 11 | 3909 | 146 | ||
| XM_011519591.2 | XP_011517893.1 | 11 | 4351 | 146 | ||
| XM_011519592.2 | XP_011517894.1 | 10 | 4543 | 146 | ||
| XM_011519593.2 | XP_011517895.1 | 10 | 4591 | 146 | ||
| XM_011519594.2 | XP_011517896.1 | 9 | 3532 | 146 | ||
| XM_011519595.2 | XP_011517897.1 | 10 | 4405 | 146 | ||
| XM_017016438.1 | XP_016871927.1 | 12 | 4209 | 146 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

CAMK1D
Genomic Map