Assay Details
| Gene Symbol: TRPC4AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015638.2 | NP_056453.1 | 10 - 11 | 1386 | 96 | |
| NM_199368.1 | NP_955400.1 | 10 - 11 | 1362 | 96 | ||
| XM_011528772.2 | XP_011527074.1 | 10 - 11 | 1408 | 96 | ||
| XM_011528774.1 | XP_011527076.1 | 5 - 6 | 699 | 96 | ||
| XM_017027799.1 | XP_016883288.1 | 10 - 11 | 1408 | 96 | ||
| XM_017027800.1 | XP_016883289.1 | 9 - 10 | 1200 | 96 | ||
| XM_017027801.1 | XP_016883290.1 | 8 - 9 | 864 | 96 | ||
| GenBank mRNA | AF055022.1 | - | 1 - 2 | 89 | 96 | |
| AK074106.1 | - | 10 - 11 | 1365 | 96 | ||
| AK298958.1 | - | 9 - 10 | 1333 | 96 | ||
| AK301368.1 | - | 2 - 3 | 305 | 96 | ||
| AK303476.1 | - | 9 - 10 | 1261 | 96 | ||
| AK315582.1 | - | 10 - 11 | 1378 | 96 | ||
| AL117480.1 | - | 4 - 5 | 642 | 96 | ||
| BC013144.1 | - | 10 - 11 | 1355 | 4475866 | 96 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000791.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20010569.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010569.hg.1 | ENST00000432634.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010569.hg.1 | ENST00000451813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010569.hg.1 | ENST00000539834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010569.hg.1 | ENST00000541994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010569.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010569.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010569.hg.1 | TC20000791.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000162357.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000162357.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000162357.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000162357.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000162357.hg.1 | TC2000008955.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000162357.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000162357.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000162357.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000162357.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000252015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | ENST00000451813.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_015638.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008955.hg.1 | NM_199368.1 | Refseq |

TRPC4AP
Genomic Map