Assay Details
| Gene Symbol: DPYSL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006426.2 | NP_006417.2 | 1 - 2 | 190 | 66 | |
| XM_011539125.2 | XP_011537427.1 | 1 - 2 | 41 | 66 | ||
| GenBank mRNA | AB006713.1 | - | 1 - 2 | 184 | 66 | |
| BC029280.1 | - | 1 - 2 | 74 | 66 | ||
| BC136329.1 | - | 1 - 2 | 59 | 9020316 | 66 | |
| BC136330.1 | - | 1 - 2 | 59 | 9020317 | 66 | |
| Y10976.1 | - | 1 - 2 | 67 | 66 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000941.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_011539125.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10011534.hg.1 | ENST00000338492.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011534.hg.1 | ENST00000368629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011534.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011534.hg.1 | TC10000941.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011537.hg.1 | ENST00000338492.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011537.hg.1 | ENST00000368629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011537.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011537.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011537.hg.1 | TC10000941.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011538.hg.1 | ENST00000338492.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011538.hg.1 | ENST00000368629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011538.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011538.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011538.hg.1 | TC10000941.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011535.hg.1 | ENST00000338492.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011535.hg.1 | ENST00000368629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011535.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011535.hg.1 | TC10000941.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000471544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000471544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000471544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164198.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164198.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164198.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164198.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164198.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164198.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164198.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164198.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164198.hg.1 | TC1000009397.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000164193.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164193.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164193.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164193.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164193.hg.1 | TC1000009397.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000164197.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164197.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164197.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164197.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164197.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164197.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164197.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164197.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164197.hg.1 | TC1000009397.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000164194.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164194.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164194.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164194.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164194.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164194.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164194.hg.1 | TC1000009397.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000471544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000471544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000471544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |

DPYSL4
Genomic Map