Assay Details
| Gene Symbol: DPYSL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011539125.2 | XP_011537427.1 | 2 - 3 | 132 | 88 | |
| XM_017015483.1 | XP_016870972.1 | 2 - 3 | 1546 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011539125.2, XM_017015483.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000471544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000471544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000471544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000338492.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000368627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000471544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000471544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000471544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | ENST00000493927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009397.hg.1 | NM_006426.2 | Refseq |

DPYSL4
Genomic Map