Assay Details
| Gene Symbol: TF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK295419.1 | - | 1 - 2 | 90 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK295419.1 transcripts.
Set Membership: |
|

TF
Genomic Map