Assay Details
| Gene Symbol: PCLO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014510.2 | NP_055325.2 | 17 - 18 | 14630 | 125 | |
| NM_033026.5 | NP_149015.2 | 17 - 18 | 14630 | 125 | ||
| XM_017012006.1 | XP_016867495.1 | 15 - 16 | 8810 | 125 | ||
| XM_017012007.1 | XP_016867496.1 | 14 - 15 | 8767 | 125 | ||
| GenBank mRNA | AB011131.1 | - | 11 - 12 | 3174 | 125 | |
| BC001304.1 | - | 8 - 9 | 836 | 125 | ||
| BC108289.1 | - | 7 - 8 | 805 | 125 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

PCLO
Genomic Map