Assay Details
| Gene Symbol: BUD31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017012760.1 | XP_016868249.1 | 1 - 2 | 103 | 80 | |
| XM_017012762.1 | XP_016868251.1 | 1 - 2 | 103 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017012760.1, XM_017012762.1 transcripts.
Set Membership: |
|

BUD31
Genomic Map