Assay Details
| Gene Symbol: XPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135669.1 | NP_001129141.1 | 3 - 4 | 419 | 129 | |
| NM_004736.3 | NP_004727.2 | 3 - 4 | 419 | 129 | ||
| GenBank mRNA | AF089744.1 | - | 3 - 4 | 392 | 129 | |
| AF099082.1 | - | 3 - 4 | 396 | 129 | ||
| AF115389.1 | - | 3 - 4 | 227 | 129 | ||
| AK309750.1 | - | 3 - 4 | 407 | 129 | ||
| BC041142.1 | - | 3 - 4 | 397 | 5582839 | 129 | |
| DB461474.1 | - | 3 - 4 | 419 | 129 | ||
More Information
Set Membership: |
|

XPR1
Genomic Map