Assay Details
| Gene Symbol: LCAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000229.1 | NP_000220.1 | 2 - 3 | 319 | 124 | |
| GenBank mRNA | BC014781.1 | - | 2 - 3 | 309 | 4155094 | 124 |
| BT009748.1 | - | 2 - 3 | 308 | 124 | ||
| M12625.1 | - | 2 - 3 | 575 | 124 | ||
| M26268.1 | - | 2 - 3 | 261 | 124 | ||
| X06537.1 | - | 2 - 3 | 411 | 124 | ||
More Information
Set Membership: |
|

LCAT
Genomic Map