Assay Details
| Gene Symbol: VTI1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006370.2 | NP_006361.1 | 2 - 3 | 573 | 146 | |
| GenBank mRNA | AF035824.1 | - | 2 - 3 | 248 | 146 | |
| AF060902.1 | - | 2 - 3 | 517 | 146 | ||
| AK316572.1 | - | 2 - 3 | 310 | 146 | ||
| BC003142.1 | - | 2 - 3 | 389 | 2958320 | 146 | |
| BT019348.1 | - | 2 - 3 | 177 | 146 | ||
| BT019960.1 | - | 2 - 3 | 146 | |||
| CR456757.1 | - | 2 - 3 | 177 | 146 | ||
| CR542095.1 | - | 2 - 3 | 177 | 146 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14002327.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein targeting to vacuole
ER to Golgi vesicle-mediated transport
intra-Golgi vesicle-mediated transport
Golgi to vacuole transport
vesicle docking involved in exocytosis
cell proliferation
vesicle-mediated transport
retrograde transport, endosome to Golgi
vesicle fusion with Golgi apparatus
membrane fusion
autophagosome maturation
regulation of protein localization to plasma membrane
SNARE binding
SNAP receptor activity
protein binding
chloride channel inhibitor activity
protein targeting to vacuole
ER to Golgi vesicle-mediated transport
intra-Golgi vesicle-mediated transport
Golgi to vacuole transport
vesicle docking involved in exocytosis
cell proliferation
vesicle-mediated transport
retrograde transport, endosome to Golgi
vesicle fusion with Golgi apparatus
membrane fusion
autophagosome maturation
regulation of protein localization to plasma membrane
SNARE binding
SNAP receptor activity
protein binding
chloride channel inhibitor activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000553619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000553619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000553619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000553619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000553619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000553619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000554636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000554636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000554636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000554636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000554636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000554636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000554636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000555543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000555543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000555543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000555543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000555543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000555543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000555543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000555543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000556461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000556461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000556461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000556461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000556461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000556461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000556461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | ENST00000556461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008877.hg.1 | ENST00000427516.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008877.hg.1 | ENST00000427516.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008877.hg.1 | ENST00000427516.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14014118.hg.1 | TC14002327.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14014118.hg.1 | ENST00000554659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14014118.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14014120.hg.1 | TC14002327.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14014120.hg.1 | ENST00000554659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14014120.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14014119.hg.1 | TC14002327.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14014119.hg.1 | ENST00000554659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14014119.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400182703.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400182703.hg.1 | ENST00000553619.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400182703.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400182703.hg.1 | ENST00000555543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400182703.hg.1 | ENST00000556461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400182703.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400182703.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400182703.hg.1 | ENST00000553619.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400182703.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400182703.hg.1 | ENST00000555543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400182703.hg.1 | ENST00000556461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400182703.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400182703.hg.1 | TC1400010764.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900008877.hg.1 | ENST00000427516.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008877.hg.1 | ENST00000427516.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008877.hg.1 | ENST00000427516.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000553619.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000553619.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000553619.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000553619.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000553619.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000553619.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000554636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000554636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000554636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000554636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000554636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000554636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000554636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000555543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000555543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000555543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000555543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000555543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000555543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000555543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000555543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000556461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000556461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000556461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000556461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000556461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000556461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000556461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | ENST00000556461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010764.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400182704.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400182704.hg.1 | ENST00000553619.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400182704.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400182704.hg.1 | ENST00000555543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400182704.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400182704.hg.1 | ENST00000216456.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400182704.hg.1 | ENST00000553619.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400182704.hg.1 | ENST00000554659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400182704.hg.1 | ENST00000555543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400182704.hg.1 | NM_006370.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400182704.hg.1 | TC1400010764.hg.1 | Clariom D TC |

VTI1B
Genomic Map