Assay Details
| Gene Symbol: AIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001624.3 | NP_001615.2 | 16 - 17 | 5116 | 70 | |
| XM_005266839.3 | XP_005266896.1 | 18 - 19 | 5954 | 70 | ||
| GenBank mRNA | AK025693.1 | - | 5 - 6 | 490 | 70 | |
| AK297008.1 | - | 6 - 7 | 676 | 70 | ||
| AK300443.1 | - | 10 - 11 | 1151 | 70 | ||
| AK316287.1 | - | 10 - 11 | 1152 | 70 | ||
| CV573838.1 | - | 5 - 6 | 528 | 70 | ||
| U83115.1 | - | 16 - 17 | 4371 | 70 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000846.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010006.hg.1 | TC06000846.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010006.hg.1 | ENST00000285105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010006.hg.1 | ENST00000369066.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010006.hg.1 | ENST00000535438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010006.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600161930.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600161930.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600161930.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600161930.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600161930.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600161930.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600161930.hg.1 | TC0600008981.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |

AIM1
Genomic Map