Assay Details
| Gene Symbol: AIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK300443.1 | - | 1 - 2 | 144 | 147 | |
| AK316287.1 | - | 1 - 2 | 145 | 147 | ||
| BC062788.1 | - | 1 - 2 | 167 | 147 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |

AIM1
Genomic Map