Assay Details
| Gene Symbol: AIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001624.3 | NP_001615.2 | 12 - 13 | 4704 | 169 | |
| XM_005266839.3 | XP_005266896.1 | 14 - 15 | 5542 | 169 | ||
| XM_017010333.1 | XP_016865822.1 | 12 - 13 | 4929 | 169 | ||
| GenBank mRNA | AK297008.1 | - | 2 - 3 | 264 | 169 | |
| AK300443.1 | - | 6 - 7 | 739 | 169 | ||
| AK316287.1 | - | 6 - 7 | 740 | 169 | ||
| BC062788.1 | - | 6 - 7 | 762 | 169 | ||
| U83115.1 | - | 12 - 13 | 3959 | 169 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000846.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06010001.hg.1 | TC06000846.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010001.hg.1 | ENST00000285105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010001.hg.1 | ENST00000369066.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010001.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010001.hg.1 | ENST00000535438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010001.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008981.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600161925.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600161925.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600161925.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600161925.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600161925.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600161925.hg.1 | ENST00000369066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600161925.hg.1 | ENST00000457437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600161925.hg.1 | ENST00000487681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600161925.hg.1 | ENST00000633556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600161925.hg.1 | NM_001624.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600161925.hg.1 | TC0600008981.hg.1 | Clariom D TC |

AIM1
Genomic Map