Assay Details
| Gene Symbol: COL19A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001858.5 | NP_001849.2 | 11 - 12 | 1143 | 104 | |
| XM_011535437.2 | XP_011533739.1 | 11 - 12 | 1410 | 104 | ||
| XM_011535438.2 | XP_011533740.1 | 11 - 12 | 1138 | 104 | ||
| XM_017010253.1 | XP_016865742.1 | 11 - 12 | 1410 | 104 | ||
| XM_017010255.1 | XP_016865744.1 | 11 - 12 | 1349 | 104 | ||
| XM_017010256.1 | XP_016865745.1 | 11 - 12 | 1143 | 104 | ||
| XM_017010257.1 | XP_016865746.1 | 10 - 11 | 1284 | 104 | ||
| XM_017010258.1 | XP_016865747.1 | 10 - 11 | 1017 | 104 | ||
| XM_017010259.1 | XP_016865748.1 | 11 - 12 | 1409 | 104 | ||
| GenBank mRNA | AK310621.1 | - | 11 - 12 | 1133 | 104 | |
| BC113362.1 | - | 11 - 12 | 1111 | 8322414 | 104 | |
| BC113364.1 | - | 11 - 12 | 1111 | 8322416 | 104 | |
| L12347.1 | - | 7 - 8 | 711 | 104 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000712.hg.1
Set Membership: |
|

COL19A1
Genomic Map