Assay Details
| Gene Symbol: COL19A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001858.5 | NP_001849.2 | 30 - 31 | 2153 | 90 | |
| XM_011535437.2 | XP_011533739.1 | 30 - 31 | 2420 | 90 | ||
| XM_011535438.2 | XP_011533740.1 | 30 - 31 | 2148 | 90 | ||
| XM_017010253.1 | XP_016865742.1 | 29 - 30 | 2378 | 90 | ||
| XM_017010254.1 | XP_016865743.1 | 29 - 30 | 2366 | 90 | ||
| XM_017010255.1 | XP_016865744.1 | 30 - 31 | 2359 | 90 | ||
| XM_017010256.1 | XP_016865745.1 | 29 - 30 | 2111 | 90 | ||
| XM_017010257.1 | XP_016865746.1 | 29 - 30 | 2294 | 90 | ||
| XM_017010258.1 | XP_016865747.1 | 29 - 30 | 2027 | 90 | ||
| XM_017010259.1 | XP_016865748.1 | 30 - 31 | 2419 | 90 | ||
| GenBank mRNA | BC113362.1 | - | 30 - 31 | 2121 | 8322414 | 90 |
| BC113364.1 | - | 30 - 31 | 2121 | 8322416 | 90 | |
| D38163.1 | - | 30 - 31 | 2153 | 90 | ||
| L12347.1 | - | 26 - 27 | 1721 | 90 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000712.hg.1
Set Membership: |
|

COL19A1
Genomic Map