Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000593604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000593604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000593604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000593604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000593604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000593604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000593604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000594398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000594398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000594398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000594398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000594398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000594398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000595886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000595886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000595886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000595886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000595886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000595886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000595886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000595886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000595886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000595886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000595886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000595886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000595886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000597578.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000597578.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000597578.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000597578.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000597578.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000597578.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000598996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000598996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000598996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000598996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000598996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000598996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000598996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000598996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000598996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000598996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000598996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000598996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000599749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000599749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000599749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000599749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000599749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000599749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000599749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000599749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000599749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000599749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000599749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000599749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000599749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000599749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000599749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000599749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000600553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000600553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000600553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000600553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000600553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
ENST00000600553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
NR_033925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
NR_033925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
NR_033925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
NR_033925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
NR_033925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
NR_033925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
NR_033925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
NR_033925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
NR_033925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
NR_033925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
NR_033925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
NR_033925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
NR_033925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
NR_036444.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
NR_036444.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
NR_036444.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
NR_036444.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
NR_036444.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
NR_036444.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
NR_036444.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011143.hg.1 |
NR_036444.1 |
Refseq |