Assay Details
| Gene Symbol: MUC16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024690.2 | NP_078966.2 | 36 - 37 | 39115 | 98 | |
| GenBank mRNA | AF361486.3 | - | 14035 | 98 | ||
| AF414442.2 | - | 39589 | 98 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001139.hg.1
Set Membership: |
|

MUC16
Genomic Map