| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015727.hg.1 |
ENST00000397910.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015727.hg.1 |
ENST00000441155.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015727.hg.1 |
NM_024690.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015727.hg.1 |
TC19001139.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015728.hg.1 |
ENST00000397910.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015728.hg.1 |
ENST00000441155.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015728.hg.1 |
NM_024690.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015728.hg.1 |
TC19001139.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900177016.hg.1 |
ENST00000397910.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900177016.hg.1 |
NM_024690.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900177016.hg.1 |
TC1900009559.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900177016.hg.1 |
ENST00000397910.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900177016.hg.1 |
NM_024690.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900177015.hg.1 |
ENST00000397910.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900177015.hg.1 |
NM_024690.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900177015.hg.1 |
TC1900009559.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900177015.hg.1 |
ENST00000397910.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900177015.hg.1 |
NM_024690.2 |
Refseq |