Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008274.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008274.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008274.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008274.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008274.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008274.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008274.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008274.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008274.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008274.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008274.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008274.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008274.hg.1 |
ENST00000437892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008274.hg.1 |
ENST00000437892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008274.hg.1 |
ENST00000437892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008274.hg.1 |
ENST00000437892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008274.hg.1 |
NR_038220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008274.hg.1 |
NR_038220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008274.hg.1 |
NR_038220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008274.hg.1 |
NR_038220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008274.hg.1 |
NR_038220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008274.hg.1 |
NR_038220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006516.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006516.hg.1 |
NR_038220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006516.hg.1 |
TC10000595.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000156086.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156086.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156086.hg.1 |
NR_038220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156086.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156086.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156086.hg.1 |
NR_038220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156086.hg.1 |
TC1000008274.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000008274.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008274.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008274.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008274.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008274.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008274.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008274.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008274.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008274.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008274.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008274.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008274.hg.1 |
ENST00000415469.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008274.hg.1 |
ENST00000437892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008274.hg.1 |
ENST00000437892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008274.hg.1 |
ENST00000437892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008274.hg.1 |
ENST00000437892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008274.hg.1 |
NR_038220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008274.hg.1 |
NR_038220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008274.hg.1 |
NR_038220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008274.hg.1 |
NR_038220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008274.hg.1 |
NR_038220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008274.hg.1 |
NR_038220.1 |
Refseq |