Assay Details
Gene Symbol: OR2M7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004691.1 | NP_001004691.1 | 1 | 879 | 148 | |
GenBank mRNA | BC136944.1 | - | 1 | 908 | 9020936 | 148 |
BC136946.1 | - | 1 | 908 | 9020938 | 148 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01004083.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931845 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006374 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15205629 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15205644 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006390 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006372 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15205666 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931858 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006415 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931851 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931859 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006361 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006373 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006414 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006371 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006384 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006380 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15205640 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006404 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931861 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006392 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006410 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15205667 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006398 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006409 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3311046 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 29122 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3331952 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3311046 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3311046 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006413 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006380 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931851 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006382 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931859 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931855 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006384 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205644 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006378 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205640 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006361 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006405 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931861 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006404 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006414 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006412 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931850 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205629 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006373 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006415 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205653 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006410 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205664 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006396 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205666 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931854 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006399 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931862 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006371 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006392 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006372 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006373 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931855 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006415 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006384 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006378 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931854 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006414 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006410 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205640 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006372 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931861 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931859 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205666 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006361 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205629 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006412 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205653 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931862 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931851 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006371 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931850 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006396 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205664 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006399 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006405 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006380 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006404 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006382 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006413 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006392 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205644 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931849 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931863 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931853 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931861 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931855 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931850 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931851 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931865 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931862 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931857 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931860 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931864 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931848 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931859 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931854 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931846 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931852 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P349716 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931852 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931850 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931864 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931846 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931859 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931861 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931848 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931856 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15205629 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931854 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931849 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15205627 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931865 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931851 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931863 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_001004691_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7925746 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7911261 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7911262 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7911260 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7925745 | - | - |
Affymetrix Human Genome U219 Array | 11738760_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11738761_PM_x_at | - | - |
Affymetrix Human Genome U219 Array | 11738761_x_at | - | - |
Affymetrix Human Genome U219 Array | 11738760_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0100018124.hg.1 | ENST00000317965.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018124.hg.1 | NM_001004691.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01061488.hg.1 | TC01004083.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01061488.hg.1 | ENST00000317965.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01061488.hg.1 | NM_001004691.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012346.hg.1 | ENST00000318104.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012346.hg.1 | ENST00000318104.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012346.hg.1 | ENST00000318104.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012346.hg.1 | NR_002141.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100236314.hg.1 | TC0100018124.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100012346.hg.1 | ENST00000318104.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012346.hg.1 | ENST00000318104.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012346.hg.1 | ENST00000318104.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012346.hg.1 | NR_002141.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018124.hg.1 | ENST00000317965.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018124.hg.1 | NM_001004691.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100236313.hg.1 | ENST00000317965.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100236313.hg.1 | NM_001004691.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100236313.hg.1 | ENST00000317965.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100236313.hg.1 | NM_001004691.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100236313.hg.1 | TC0100018124.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100236312.hg.1 | TC0100018124.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A013740 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A013740 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 131980 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1655713 | BC136946.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1655713 | NM_001004691.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1655713 | BC136944.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1655713 | BC136946.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1655713 | NM_001004691.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1655713 | BC136944.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1655713 | BC136946.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1655713 | NM_001004691.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1655713 | BC136944.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1655713 | BC136946.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1655713 | NM_001004691.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1655713 | BC136944.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1655713 | BC136946.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1655713 | NM_001004691.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1655713 | BC136944.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_23714-S | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1655713 | BC136946.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1655713 | NM_001004691.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1655713 | BC136944.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1655713 | BC136946.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1655713 | NM_001004691.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1655713 | BC136944.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 6290156 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_23714 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1655713 | BC136946.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1655713 | NM_001004691.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1655713 | BC136944.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1655713 | BC136946.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1655713 | NM_001004691.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1655713 | BC136944.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1655713 | BC136946.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1655713 | NM_001004691.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1655713 | BC136944.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1655713 | BC136946.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1655713 | NM_001004691.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1655713 | BC136944.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1655713 | BC136946.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1655713 | NM_001004691.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1655713 | BC136944.1 | human_transcriptome |
