Assay Details
| Gene Symbol: HTR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000863.2 | NP_000854.1 | 1 | 1756 | 144 | |
| GenBank mRNA | AK290080.1 | - | 1 | 1507 | 144 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

HTR1B
Genomic Map