Assay Details
| Gene Symbol: ORC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190818.1 | NP_001177747.1 | 7 - 8 | 1414 | 115 | |
| NM_001190819.1 | NP_001177748.1 | 7 - 8 | 1417 | 115 | ||
| NM_004153.3 | NP_004144.2 | 7 - 8 | 1417 | 115 | ||
| XM_011541527.2 | XP_011539829.1 | 7 - 8 | 1366 | 115 | ||
| XM_017001388.1 | XP_016856877.1 | 7 - 8 | 1417 | 115 | ||
| XM_017001389.1 | XP_016856878.1 | 8 - 9 | 1454 | 115 | ||
| GenBank mRNA | AK291909.1 | - | 7 - 8 | 1392 | 115 | |
| AK303420.1 | - | 7 - 8 | 1358 | 115 | ||
| BC011539.1 | - | 7 - 8 | 1330 | 3870637 | 115 | |
| U40152.1 | - | 7 - 8 | 1405 | 115 | ||
| U43416.1 | - | 7 - 8 | 1397 | 115 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002658.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
G1/S transition of mitotic cell cycle
regulation of transcription involved in G1/S transition of mitotic cell cycle
DNA replication
DNA replication initiation
positive regulation of smooth muscle cell proliferation
positive regulation of G0 to G1 transition
DNA binding
chromatin binding
protein binding
ATP binding
G1/S transition of mitotic cell cycle
regulation of transcription involved in G1/S transition of mitotic cell cycle
DNA replication
DNA replication initiation
positive regulation of smooth muscle cell proliferation
positive regulation of G0 to G1 transition
DNA binding
chromatin binding
protein binding
ATP binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041682.hg.1 | TC01002658.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041682.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041682.hg.1 | ENST00000371568.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041682.hg.1 | ENST00000476332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041682.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041682.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041682.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014202.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206206.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206206.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206206.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206206.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206206.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206206.hg.1 | ENST00000371566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206206.hg.1 | ENST00000371568.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206206.hg.1 | NM_001190818.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206206.hg.1 | NM_001190819.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206206.hg.1 | NM_004153.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206206.hg.1 | TC0100014202.hg.1 | Clariom D TC |

ORC1
Genomic Map