| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05019793.hg.1 |
TC05001419.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05019793.hg.1 |
ENST00000256447.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05019793.hg.1 |
NM_005582.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05019796.hg.1 |
TC05001419.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05019796.hg.1 |
ENST00000256447.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05019796.hg.1 |
NM_005582.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05019795.hg.1 |
TC05001419.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05019795.hg.1 |
ENST00000256447.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05019795.hg.1 |
NM_005582.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013316.hg.1 |
ENST00000515027.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013316.hg.1 |
ENST00000515027.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013316.hg.1 |
ENST00000515027.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013316.hg.1 |
NM_005582.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013316.hg.1 |
NM_005582.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013316.hg.1 |
NM_005582.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013316.hg.1 |
NM_005582.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013316.hg.1 |
NM_005582.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013316.hg.1 |
NM_005582.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013316.hg.1 |
NM_005582.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013316.hg.1 |
NM_005582.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500195669.hg.1 |
NM_005582.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500195669.hg.1 |
NM_005582.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500195669.hg.1 |
TC0500013316.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500195666.hg.1 |
ENST00000515027.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500195666.hg.1 |
NM_005582.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500195666.hg.1 |
ENST00000515027.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500195666.hg.1 |
NM_005582.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500195666.hg.1 |
TC0500013316.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500195668.hg.1 |
NM_005582.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500195668.hg.1 |
NM_005582.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500195668.hg.1 |
TC0500013316.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500195670.hg.1 |
NM_005582.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500195670.hg.1 |
NM_005582.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500195670.hg.1 |
TC0500013316.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500013316.hg.1 |
ENST00000515027.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013316.hg.1 |
ENST00000515027.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013316.hg.1 |
ENST00000515027.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013316.hg.1 |
NM_005582.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013316.hg.1 |
NM_005582.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013316.hg.1 |
NM_005582.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013316.hg.1 |
NM_005582.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013316.hg.1 |
NM_005582.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013316.hg.1 |
NM_005582.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013316.hg.1 |
NM_005582.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500013316.hg.1 |
NM_005582.2 |
Refseq |