| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01018448.hg.1 |
TC01001197.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01018448.hg.1 |
ENST00000368947.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01018448.hg.1 |
ENST00000474997.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01018448.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
ENST00000474997.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
ENST00000474997.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
ENST00000474997.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
ENST00000474997.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
ENST00000474997.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
ENST00000474997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
ENST00000474997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
ENST00000474997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
ENST00000474997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
ENST00000474997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009934.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100171803.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100171803.hg.1 |
ENST00000474997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100171803.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100171803.hg.1 |
ENST00000368947.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100171803.hg.1 |
ENST00000474997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100171803.hg.1 |
NM_003568.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100171803.hg.1 |
TC0100009934.hg.1 |
Clariom D TC |