Assay Details
Gene Symbol: TMPRSS11E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014058.3 | NP_054777.2 | 9 - 10 | 1179 | 102 | |
XM_011531896.2 | XP_011530198.1 | 8 - 9 | 1029 | 102 | ||
GenBank mRNA | AF064819.1 | - | 9 - 10 | 1167 | 102 | |
AK313859.1 | - | 9 - 10 | 1179 | 102 | ||
AY359017.1 | - | 9 - 10 | 1146 | 102 | ||
BC113412.1 | - | 9 - 10 | 1138 | 8322464 | 102 | |
BC113414.1 | - | 9 - 10 | 1138 | 8322466 | 102 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04000369.hg.1, TC4_ctg9_hap1000001.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P18751 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P18751 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 8735 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 30389 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P18751 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P18751 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23319319 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23319319 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P18751 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P18751 | - | - |
Agilent Human 2 cDNA Microarray (G4101A) | 2662067 | - | - |
Agilent Human 2 cDNA Microarray (G4101A) | 2662067 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P454048 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8095383 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8095388 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8095372 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8095369 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8095382 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8095373 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8095368 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8095387 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8095381 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8095389 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8095375 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8095385 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8095386 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8095374 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8095365 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8095367 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8095366 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8095370 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8095384 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8095371 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8095364 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8095380 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 220431_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11736715_PM_s_at | - | - |
Affymetrix Human Genome U219 Array | 11736715_s_at | - | - |
Affymetrix Human Genome U219 Array | 11736716_PM_s_at | - | - |
Affymetrix Human Genome U219 Array | 11736716_s_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR4_ctg9_hap1000012.hg.1 | TC4_ctg9_hap1000001.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR4_ctg9_hap1000012.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | ENST00000510647.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | ENST00000510647.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | ENST00000510647.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | ENST00000510647.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | ENST00000510647.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | ENST00000510647.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007734.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR4_ctg9_hap1000011.hg.1 | TC4_ctg9_hap1000001.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR4_ctg9_hap1000011.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04005008.hg.1 | TC04000369.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04005008.hg.1 | ENST00000305363.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005008.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04005009.hg.1 | TC04000369.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04005009.hg.1 | ENST00000305363.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005009.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | ENST00000510647.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | ENST00000510647.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | ENST00000510647.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | ENST00000510647.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | ENST00000510647.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | ENST00000510647.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007734.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400152705.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400152705.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400152705.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400152705.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400152705.hg.1 | TC0400007734.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400152706.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400152706.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400152706.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400152706.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400152706.hg.1 | TC0400007734.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400152704.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400152704.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400152704.hg.1 | ENST00000305363.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400152704.hg.1 | NM_014058.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400152704.hg.1 | TC0400007734.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 220431_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 220431_at | - | - |
Human Genome U133A 2.0 Array | 220431_PM_at | - | - |
Human Genome U133A 2.0 Array | 220431_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw569246 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220431_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220431_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220431_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220431_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220431_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220431_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220431_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220431_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220431_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220431_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220431_at10 | - | - |
Human Genome U95 Set | 69149_PM_at | - | - |
Human Genome U95 Set | 69149_at | - | - |
Human Genome U133 Set | 220431_at | - | - |
Human Genome U133 Set | 220431_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A025604 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A025604 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 129331 | - | - |
ABI Human Genome Survey Microarray Version 2.0 | 137941 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1740007 | AY359017.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1740007 | NM_014058.3 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1740007 | AY359017.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1740007 | NM_014058.3 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1740007 | AY359017.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1740007 | NM_014058.3 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_40254870-S | BC113414.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_40254870-S | BC113412.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_40254870-S | AK292787.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_40254870-S | AK313859.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_40254870-S | AY359017.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_25694-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0005720288 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0005720288 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1740007 | AY359017.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1740007 | NM_014058.3 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1740007 | AY359017.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1740007 | NM_014058.3 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 3780341 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_25694 | - | - |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1740007 | AY359017.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1740007 | NM_014058.3 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1740007 | AY359017.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1740007 | NM_014058.3 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1740007 | AY359017.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1740007 | NM_014058.3 | human_transcriptome |
