Assay Details
| Gene Symbol: TBCCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018138.4 | NP_060608.1 | 1 - 2 | 92 | 58 | |
| GenBank mRNA | AK001422.1 | - | 1 - 2 | 65 | 58 | |
| BC025748.1 | - | 1 - 2 | 78 | 5200082 | 58 | |
| DB023715.1 | - | 1 - 2 | 90 | 58 | ||
| HY017180.1 | - | 1 - 2 | 92 | 58 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03002084.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK001422.1,BC025748.1,NM_018138.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03037047.hg.1 | TC03002084.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037047.hg.1 | ENST00000338733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037047.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037046.hg.1 | TC03002084.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037046.hg.1 | ENST00000338733.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037046.hg.1 | ENST00000424280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037046.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037046.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000413695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000413695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000413695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000413695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000413695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000413695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000413695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000413695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000430560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000430560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000446782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000446782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000446782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000446782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000446782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000446782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000446782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000446782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000446782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000446782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000446782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000479590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | ENST00000479590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001286749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001286749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001286749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001286749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001286749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001286749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001286749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001286749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001286749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001286749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001286749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001286749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_001286749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014085.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000413695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000413695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000413695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000413695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000413695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000413695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000413695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000413695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000430560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000430560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000446782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000446782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000446782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000446782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000446782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000446782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000446782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000446782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000446782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000446782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000446782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000479590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | ENST00000479590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001286749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001286749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001286749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001286749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001286749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001286749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001286749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001286749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001286749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001286749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001286749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001286749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_001286749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014085.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300209788.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209788.hg.1 | ENST00000413695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209788.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209788.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300209788.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300209788.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209788.hg.1 | ENST00000413695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209788.hg.1 | ENST00000424280.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209788.hg.1 | NM_001134415.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300209788.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300209788.hg.1 | TC0300014085.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300209789.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209789.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300209789.hg.1 | ENST00000338733.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209789.hg.1 | NM_018138.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300209789.hg.1 | TC0300014085.hg.1 | Clariom D TC |

TBCCD1
Genomic Map