Assay Details
| Gene Symbol: GLMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256605.1 | NP_001243534.1 | 3 - 4 | 582 | 71 | |
| NM_001256609.1 | NP_001243538.1 | 4 - 5 | 1069 | 71 | ||
| NM_144580.2 | NP_653181.1 | 4 - 5 | 840 | 71 | ||
| XM_011509117.2 | XP_011507419.1 | 3 - 4 | 499 | 71 | ||
| GenBank mRNA | AK075349.1 | - | 4 - 5 | 809 | 71 | |
| AK296157.1 | - | 4 - 5 | 1037 | 71 | ||
| AY358450.1 | - | 4 - 5 | 813 | 71 | ||
| BC011575.1 | - | 4 - 5 | 823 | 71 | ||
| BC018757.1 | - | 4 - 5 | 809 | 4850051 | 71 | |
| BC036340.1 | - | 4 - 5 | 756 | 71 | ||
| BM833126.1 | - | 2 - 3 | 242 | 71 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003335.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
intracellular receptor signaling pathway
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, sequence-specific DNA binding
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
ligand-dependent nuclear receptor transcription coactivator activity
transcription regulatory region DNA binding
intracellular receptor signaling pathway
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, sequence-specific DNA binding
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
ligand-dependent nuclear receptor transcription coactivator activity
transcription regulatory region DNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000362007.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000362007.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000362007.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000362007.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000362007.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000362007.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000362007.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000362007.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000362007.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000362007.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000362007.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000362007.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000362007.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000362007.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000362007.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000368264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000368264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000368264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000368264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000368264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000368264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000368264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000368264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000368264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000368264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000476177.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000476177.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000476177.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000476177.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000476177.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000476177.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000476177.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000476177.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000479084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000479084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000479084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000479084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000479084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000479084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000479084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000479084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000479084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000481050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000481050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000481050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000481050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000481050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000481050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000481050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000482579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000482579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000482579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000482579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000482579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000484214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000484214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000484214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000484214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000484214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000484214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000484214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000484214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000497955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000497955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000497955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000497955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000497955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000497955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000497955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000497955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000497955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000497955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000612353.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000612353.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000612353.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000612353.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000612353.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000612353.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000612353.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000612353.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000612353.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000612353.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000612353.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000614643.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000614643.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000614643.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000614643.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000614643.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000614643.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000614643.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000614643.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000614643.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000614643.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000614643.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000614643.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000614643.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000614643.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000614643.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000614643.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000622703.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000622703.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000622703.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000622703.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000622703.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000622703.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000622703.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000622703.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000622703.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000622703.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000622703.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000622703.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | ENST00000622703.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_001256609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_144580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_144580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_144580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_144580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_144580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_144580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_144580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_144580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_144580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_144580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_144580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_144580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_144580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_144580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018510.hg.1 | NM_144580.2 | Refseq |

GLMP
Genomic Map