| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07021024.hg.1 |
TC07001348.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07021024.hg.1 |
ENST00000258787.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07021024.hg.1 |
NM_033054.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700178165.hg.1 |
ENST00000258787.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700178165.hg.1 |
ENST00000463516.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700178165.hg.1 |
ENST00000483585.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700178165.hg.1 |
ENST00000488554.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700178165.hg.1 |
ENST00000495831.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700178165.hg.1 |
NM_033054.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700178165.hg.1 |
ENST00000258787.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700178165.hg.1 |
ENST00000463516.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700178165.hg.1 |
ENST00000483585.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700178165.hg.1 |
ENST00000488554.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700178165.hg.1 |
ENST00000495831.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700178165.hg.1 |
NM_033054.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700178165.hg.1 |
TC0700010941.hg.1 |
Clariom D TC |