Assay Details
| Gene Symbol: MAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032272.4 | NP_115648.2 | 5 - 6 | 1002 | 81 | |
| XM_017013903.1 | XP_016869392.1 | 5 - 6 | 1125 | 81 | ||
| XM_017013904.1 | XP_016869393.1 | 5 - 6 | 711 | 81 | ||
| GenBank mRNA | AL136937.1 | - | 5 - 6 | 893 | 81 | |
| BC014082.2 | - | 5 - 6 | 894 | 4304506 | 81 | |
| BC018714.1 | - | 5 - 6 | 901 | 5014952 | 81 | |
| BC031273.1 | - | 5 - 6 | 904 | 5288176 | 81 | |
| BG757465.1 | - | 3 - 4 | 434 | 81 | ||
| CR533463.1 | - | 4 - 5 | 501 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000841.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
negative regulation of transcription from RNA polymerase III promoter
transcription factor TFIIIB complex assembly
RNA polymerase III core binding
RNA polymerase III type 1 promoter DNA binding
RNA polymerase III type 2 promoter DNA binding
RNA polymerase III type 3 promoter DNA binding
GABA receptor binding
transcription, DNA-templated
negative regulation of transcription from RNA polymerase III promoter
transcription factor TFIIIB complex assembly
RNA polymerase III core binding
RNA polymerase III type 1 promoter DNA binding
RNA polymerase III type 2 promoter DNA binding
RNA polymerase III type 3 promoter DNA binding
GABA receptor binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000527058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000527058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000527058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000527058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000527058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000527058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000527058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000527058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000527058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000527058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000527572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000527572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000527572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000527572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000527572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000527572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000534811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000534811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000534811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000534811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000534811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | ENST00000534811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009240.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011129.hg.1 | TC08000841.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011129.hg.1 | ENST00000322428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011129.hg.1 | ENST00000532522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011129.hg.1 | ENST00000534585.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011129.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011131.hg.1 | TC08000841.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011131.hg.1 | ENST00000322428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011131.hg.1 | ENST00000532522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011131.hg.1 | ENST00000534585.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011131.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163106.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163106.hg.1 | ENST00000527058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163106.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163106.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163106.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163106.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163106.hg.1 | ENST00000527058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163106.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163106.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163106.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163106.hg.1 | TC0800009240.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800163108.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163108.hg.1 | ENST00000527058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163108.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163108.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163108.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163108.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163108.hg.1 | ENST00000527058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163108.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163108.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163108.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163108.hg.1 | TC0800009240.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800163109.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163109.hg.1 | ENST00000527058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163109.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163109.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163109.hg.1 | ENST00000534811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163109.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163109.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163109.hg.1 | ENST00000527058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163109.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163109.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163109.hg.1 | ENST00000534811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163109.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163109.hg.1 | TC0800009240.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000322428.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000527058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000527058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000527058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000527058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000527058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000527058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000527058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000527058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000527058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000527058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000527572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000527572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000527572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000527572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000527572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000527572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000532522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000534585.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000534811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000534811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000534811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000534811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000534811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | ENST00000534811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | NM_032272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009240.hg.1 | NM_032272.4 | Refseq |

MAF1
Genomic Map