Assay Details
| Gene Symbol: CHIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012110.3 | NP_036242.1 | 4 - 5 | 728 | 101 | |
| GenBank mRNA | AF159423.1 | - | 4 - 5 | 629 | 101 | |
| AK312427.1 | - | 4 - 5 | 691 | 101 | ||
| BC034691.1 | - | 4 - 5 | 640 | 4747392 | 101 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001201.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000509678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000509678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000509678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000509678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000509678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000510894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000510894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000510894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000510894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000510894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000510894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000510894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000512964.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000512964.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000512964.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000512964.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000512964.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000512964.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000512964.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | ENST00000512964.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | NM_012110.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | NM_012110.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | NM_012110.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | NM_012110.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | NM_012110.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | NM_012110.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | NM_012110.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | NM_012110.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | NM_012110.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | NM_012110.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | NM_012110.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010715.hg.1 | NM_012110.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04016478.hg.1 | TC04001201.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04016478.hg.1 | ENST00000263921.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04016478.hg.1 | ENST00000512964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04016478.hg.1 | NM_012110.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400172940.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400172940.hg.1 | ENST00000510894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400172940.hg.1 | ENST00000512964.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400172940.hg.1 | NM_012110.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400172940.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400172940.hg.1 | ENST00000510894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400172940.hg.1 | ENST00000512964.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400172940.hg.1 | NM_012110.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400172940.hg.1 | TC0400010715.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000263921.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000509678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000509678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000509678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000509678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000509678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000510894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000510894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000510894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000510894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000510894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000510894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000510894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000512964.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000512964.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000512964.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000512964.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000512964.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000512964.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000512964.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | ENST00000512964.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | NM_012110.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | NM_012110.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | NM_012110.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | NM_012110.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | NM_012110.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | NM_012110.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | NM_012110.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | NM_012110.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | NM_012110.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | NM_012110.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | NM_012110.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010715.hg.1 | NM_012110.3 | Refseq |

CHIC2
Genomic Map