Assay Details
| Gene Symbol: PAX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257096.1 | NP_001244025.1 | 3 - 4 | 1118 | 72 | |
| NM_006192.4 | NP_006183.2 | 3 - 4 | 1118 | 72 | ||
| GenBank mRNA | AK303335.1 | - | 2 - 3 | 999 | 72 | |
| BC069134.1 | - | 2 - 3 | 782 | 7216990 | 72 | |
| BC143791.1 | - | 3 - 4 | 866 | 72 | ||
| BC143793.1 | - | 3 - 4 | 863 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000145.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
somitogenesis
transcription from RNA polymerase II promoter
cell proliferation
CD4-positive, alpha-beta T cell differentiation
CD8-positive, alpha-beta T cell differentiation
positive regulation of transcription from RNA polymerase II promoter
thymus development
parathyroid gland development
bone morphogenesis
sclerotome development
DNA binding
transcription factor activity, sequence-specific DNA binding
skeletal system development
somitogenesis
transcription from RNA polymerase II promoter
cell proliferation
CD4-positive, alpha-beta T cell differentiation
CD8-positive, alpha-beta T cell differentiation
positive regulation of transcription from RNA polymerase II promoter
thymus development
parathyroid gland development
bone morphogenesis
sclerotome development
DNA binding
transcription factor activity, sequence-specific DNA binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20001917.hg.1 | ENST00000398485.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001917.hg.1 | ENST00000444366.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001917.hg.1 | ENST00000460221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001917.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001917.hg.1 | TC20000145.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001916.hg.1 | ENST00000398485.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001916.hg.1 | ENST00000444366.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001916.hg.1 | ENST00000460221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001916.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001916.hg.1 | TC20000145.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000444366.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000444366.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000444366.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000444366.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000444366.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000444366.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000444366.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000444366.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000444366.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000460221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000460221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000460221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000460221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000460221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000460221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000485038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000485038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000485038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000444366.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000444366.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000444366.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000444366.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000444366.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000444366.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000444366.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000444366.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000444366.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000460221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000460221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000460221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000460221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000460221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000460221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000485038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000485038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000485038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006913.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000147338.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000147338.hg.1 | ENST00000444366.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000147338.hg.1 | ENST00000460221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000147338.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000147338.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000147338.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000147338.hg.1 | TC2000006913.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000147338.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000147338.hg.1 | ENST00000444366.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000147338.hg.1 | ENST00000460221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000147338.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000147338.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000147338.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000147337.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000147337.hg.1 | ENST00000444366.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000147337.hg.1 | ENST00000460221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000147337.hg.1 | ENST00000485038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000147337.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000147337.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000147337.hg.1 | NM_006192.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000147337.hg.1 | TC2000006913.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000147337.hg.1 | ENST00000398485.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000147337.hg.1 | ENST00000444366.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000147337.hg.1 | ENST00000460221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000147337.hg.1 | ENST00000485038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000147337.hg.1 | ENST00000613128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000147337.hg.1 | NM_001257096.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000147337.hg.1 | NM_006192.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 178411 | - | - |

PAX1
Genomic Map