Assay Details
| Gene Symbol: GOLGA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005113.3 | NP_005104.3 | 1 - 2 | 224 | 137 | |
| XM_011537420.2 | XP_011535722.1 | 1 - 2 | 224 | 137 | ||
| GenBank mRNA | AK293077.1 | - | 1 - 2 | 127 | 137 | |
| AK303789.1 | - | 1 - 2 | 130 | 137 | ||
| BC023021.1 | - | 1 - 2 | 132 | 4999710 | 137 | |
| DB096772.1 | - | 1 - 2 | 224 | 137 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000572.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK293077.1,AK303789.1,BC023021.1,NM_005113.3,XM_011537420.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14006977.hg.1 | ENST00000163416.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006977.hg.1 | ENST00000355976.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006977.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006977.hg.1 | TC14000572.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | ENST00000554700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | ENST00000554700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | ENST00000554700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | ENST00000554700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | ENST00000555793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | ENST00000555793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | ENST00000555793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006975.hg.1 | ENST00000163416.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006975.hg.1 | ENST00000355976.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006975.hg.1 | ENST00000439315.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006975.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006975.hg.1 | TC14000572.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006976.hg.1 | ENST00000163416.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006976.hg.1 | ENST00000355976.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006976.hg.1 | ENST00000439315.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006976.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006976.hg.1 | TC14000572.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010554.hg.1 | ENST00000507317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010554.hg.1 | ENST00000507317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156762.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156762.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156762.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156762.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156762.hg.1 | TC1400008019.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400156764.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156764.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156764.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156764.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156764.hg.1 | TC1400008019.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400156766.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156766.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156766.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156766.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156766.hg.1 | TC1400008019.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400156765.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156765.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156765.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156765.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156765.hg.1 | TC1400008019.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500010554.hg.1 | ENST00000507317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010554.hg.1 | ENST00000507317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | ENST00000163416.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | ENST00000554700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | ENST00000554700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | ENST00000554700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | ENST00000554700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | ENST00000555793.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | ENST00000555793.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | ENST00000555793.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008019.hg.1 | NM_005113.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 119103 | - | - |

GOLGA5
Genomic Map