Assay Details
| Gene Symbol: ETNK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018638.4 | NP_061108.2 | 2 - 3 | 703 | 89 | |
| XM_017019580.1 | XP_016875069.1 | 2 - 3 | 700 | 89 | ||
| XM_017019581.1 | XP_016875070.1 | 2 - 3 | 700 | 89 | ||
| GenBank mRNA | AF207600.2 | - | 2 - 3 | 769 | 89 | |
| AK074425.1 | - | 2 - 3 | 648 | 89 | ||
| AL137750.1 | - | 2 - 3 | 652 | 89 | ||
| BC066907.1 | - | 2 - 3 | 703 | 5260109 | 89 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000239.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000539974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000539974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000539974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000544191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000544191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000544191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000544191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000545979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000545979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002941.hg.1 | ENST00000266517.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002941.hg.1 | ENST00000381409.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002941.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002941.hg.1 | TC12000239.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012012.hg.1 | ENST00000506341.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012012.hg.1 | ENST00000506341.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000539974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000539974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000539974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000544191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000544191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000544191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000544191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000545979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000545979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200149573.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200149573.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200149573.hg.1 | ENST00000539974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200149573.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200149573.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200149573.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200149573.hg.1 | ENST00000539974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200149573.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200149573.hg.1 | TC1200007070.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400012012.hg.1 | ENST00000506341.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012012.hg.1 | ENST00000506341.2 | Ensembl |

ETNK1
Genomic Map