Assay Details
| Gene Symbol: XDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000379.3 | NP_000370.2 | 24 - 25 | 2716 | 92 | |
| XM_011533095.2 | XP_011531397.1 | 24 - 25 | 2745 | 92 | ||
| XM_011533096.2 | XP_011531398.1 | 24 - 25 | 2747 | 92 | ||
| GenBank mRNA | D11456.2 | - | 24 - 25 | 2694 | 92 | |
| U06117.1 | - | 24 - 25 | 2700 | 92 | ||
| U39487.1 | - | 24 - 25 | 2671 | 92 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001714.hg.1
Set Membership: |
|

XDH
Genomic Map