Assay Details
| Gene Symbol: GPATCH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018040.3 | NP_060510.1 | 5 - 6 | 1223 | 83 | |
| XM_011509689.2 | XP_011507991.1 | 5 - 6 | 1433 | 83 | ||
| XM_011509690.2 | XP_011507992.1 | 5 - 6 | 1433 | 83 | ||
| XM_011509691.2 | XP_011507993.1 | 4 - 5 | 1250 | 83 | ||
| XM_011509693.2 | XP_011507995.1 | 5 - 6 | 1433 | 83 | ||
| XM_017001592.1 | XP_016857081.1 | 5 - 6 | 1433 | 83 | ||
| XM_017001593.1 | XP_016857082.1 | 5 - 6 | 1433 | 83 | ||
| GenBank mRNA | AK001114.1 | - | 5 - 6 | 1190 | 83 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003823.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01058470.hg.1 | TC01003823.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01058470.hg.1 | ENST00000366935.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01058470.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000485274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000485274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000485274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000485274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000485274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000485274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000485274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000485274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100231171.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100231171.hg.1 | ENST00000470014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100231171.hg.1 | ENST00000485274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100231171.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100231171.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100231171.hg.1 | ENST00000470014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100231171.hg.1 | ENST00000485274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100231171.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100231171.hg.1 | TC0100017321.hg.1 | Clariom D TC |

GPATCH2
Genomic Map