Assay Details
| Gene Symbol: GPATCH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018040.3 | NP_060510.1 | 8 - 9 | 1412 | 114 | |
| XM_011509689.2 | XP_011507991.1 | 8 - 9 | 1622 | 114 | ||
| XM_011509691.2 | XP_011507993.1 | 7 - 8 | 1439 | 114 | ||
| XM_011509693.2 | XP_011507995.1 | 8 - 9 | 1622 | 114 | ||
| GenBank mRNA | AK001114.1 | - | 8 - 9 | 1379 | 114 | |
| AK310304.1 | - | 4 - 5 | 692 | 114 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003823.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01058460.hg.1 | TC01003823.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01058460.hg.1 | ENST00000366935.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01058460.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000485274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000485274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000485274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000485274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100231159.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100231159.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100231159.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100231159.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100231159.hg.1 | TC0100017321.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366934.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000366935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000470014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000485274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000485274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000485274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000485274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | ENST00000489246.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_001297754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017321.hg.1 | NM_018040.4 | Refseq |

GPATCH2
Genomic Map