Assay Details
| Gene Symbol: TRHDE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013381.2 | NP_037513.1 | 1 - 2 | 811 | 74 | |
| XM_005268819.4 | XP_005268876.1 | 1 - 2 | 916 | 74 | ||
| XM_017019243.1 | XP_016874732.1 | 1 - 2 | 916 | 74 | ||
| GenBank mRNA | AF126372.1 | - | 1 - 2 | 877 | 74 | |
| AY358765.1 | - | 1 - 2 | 804 | 74 | ||
| BC142706.1 | - | 1 - 2 | 811 | 40148814 | 74 | |
| BC150181.1 | - | 1 - 2 | 811 | 40148096 | 74 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000630.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000547300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000547300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000547300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000547300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000547300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549138.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549138.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549138.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549138.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549138.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549138.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549138.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549138.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549138.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000552503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000552503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000552503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000552503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000552503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000435350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000435350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000435350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000435350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000435350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000435350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000435350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000435350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000435350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000435350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000435350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000435350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000549957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000549957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000549957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000549957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000549957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000549957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000549957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000549957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000549957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000549957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000550334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000550334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000550334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000550334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000550334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |

TRHDE
Genomic Map