Assay Details
| Gene Symbol: TBCE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079515.2 | NP_001072983.1 | 3 - 4 | 411 | 127 | |
| NM_001287801.1 | NP_001274730.1 | 3 - 4 | 363 | 127 | ||
| NM_003193.4 | NP_003184.1 | 3 - 4 | 363 | 127 | ||
| GenBank mRNA | AK292287.1 | - | 3 - 4 | 268 | 127 | |
| AK296185.1 | - | 3 - 4 | 293 | 127 | ||
| AK298050.1 | - | 3 - 4 | 293 | 127 | ||
| BC008654.1 | - | 3 - 4 | 255 | 3872809 | 127 | |
| BT007086.1 | - | 2 - 3 | 187 | 127 | ||
| CB110473.1 | - | 3 - 4 | 363 | 127 | ||
| DA418943.1 | - | 3 - 4 | 310 | 127 | ||
| DA981183.1 | - | 3 - 4 | 358 | 127 | ||
| U61232.1 | - | 3 - 4 | 316 | 127 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK298050.1 transcripts.
Set Membership: |
|

TBCE
Genomic Map