Assay Details
| Gene Symbol: ATP6V0B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001294333.1 | NP_001281262.1 | 2 - 3 | 509 | 63 | |
| NM_004047.4 | NP_004038.1 | 2 - 3 | 509 | 63 | ||
| GenBank mRNA | BC000423.2 | - | 2 - 3 | 180 | 2964510 | 63 |
| BC005876.2 | - | 2 - 3 | 180 | 2964510 | 63 | |
| BG831921.1 | - | 2 - 3 | 251 | 63 | ||
| BT007151.1 | - | 2 - 3 | 116 | 63 | ||
| CR456972.1 | - | 2 - 3 | 116 | 63 | ||
| D89052.1 | - | 2 - 3 | 198 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
vacuolar acidification
insulin receptor signaling pathway
ATP hydrolysis coupled proton transport
proton transport
regulation of macroautophagy
transferrin transport
ion transmembrane transport
phagosome acidification
transporter activity
proton-transporting ATPase activity, rotational mechanism
vacuolar acidification
insulin receptor signaling pathway
ATP hydrolysis coupled proton transport
proton transport
regulation of macroautophagy
transferrin transport
ion transmembrane transport
phagosome acidification
transporter activity
proton-transporting ATPase activity, rotational mechanism
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000236067.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000236067.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000236067.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000236067.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000236067.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000236067.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000236067.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000236067.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000236067.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000236067.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000236067.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000236067.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000236067.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000461670.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000461670.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000461670.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000461670.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000461670.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000461670.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000461670.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000461670.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000468183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000468183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000468183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000468183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000468183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000468183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000468183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000468183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000468183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000468183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000468183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000468183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000468183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000468183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000468183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000468183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000468183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000468183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000468183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000471859.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000471859.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000471859.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000471859.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000471859.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000471859.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000471859.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000471859.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000471859.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000471859.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000471859.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000471859.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472174.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472174.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472174.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472174.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472174.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472174.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472174.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472174.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472174.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472174.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472174.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472174.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472174.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472174.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472174.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472505.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472505.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472505.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472505.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472505.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472505.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000472505.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000473485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000473485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000473485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000473485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000473485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000473485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000473485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000473485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000473485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000473485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000473485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000473485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000473485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000473485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000473485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000473485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000473485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000496131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000496131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000496131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000496131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000496131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000496131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000496131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000496131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000496131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000496131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000496131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000496131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000496131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000498208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000498208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000498208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000498208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000498208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000498664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000498664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000498664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000498664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000498664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000498664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000498664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000498664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000498664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000498664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000498664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000498664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000532072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000532072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000532072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000532072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000532072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000532072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000532642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000532642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000532642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000532642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000532642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000532642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000532642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000532642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000532642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000532642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000532642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000532642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000532642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000532642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | ENST00000532642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001039457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001039457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001039457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001039457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001039457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001039457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001039457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001039457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001039457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001039457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001039457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001039457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001039457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001039457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001294333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001294333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001294333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001294333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001294333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001294333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001294333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001294333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001294333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001294333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001294333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001294333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001294333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001294333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001294333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001294333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_001294333.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_004047.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_004047.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_004047.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_004047.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_004047.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_004047.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_004047.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_004047.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_004047.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_004047.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_004047.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_004047.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_004047.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_004047.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008081.hg.1 | NM_004047.4 | Refseq |

ATP6V0B
Genomic Map