Assay Details
| Gene Symbol: ATN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007026.1 | NP_001007027.1 | 8 - 9 | 3597 | 109 | |
| NM_001940.3 | NP_001931.2 | 8 - 9 | 3590 | 109 | ||
| GenBank mRNA | BC051795.2 | - | 8 - 9 | 3611 | 4181592 | 109 |
| D31840.1 | - | 8 - 9 | 3583 | 109 | ||
| D38529.1 | - | 7 - 8 | 3504 | 109 | ||
| U23851.1 | - | 7 - 8 | 3415 | 109 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12003288.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000537488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000537488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000537488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000537488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000537488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000537488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000541029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000541029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | ENST00000541029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001298.hg.1 | TC12003288.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001298.hg.1 | ENST00000229279.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001298.hg.1 | ENST00000356654.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001298.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001298.hg.1 | ENST00000544325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001298.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001298.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200146335.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200146335.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200146335.hg.1 | ENST00000537488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200146335.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200146335.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200146335.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200146335.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200146335.hg.1 | ENST00000537488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200146335.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200146335.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200146335.hg.1 | TC1200006654.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000356654.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000396684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000537488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000537488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000537488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000537488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000537488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000537488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000541029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000541029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | ENST00000541029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001007026.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006654.hg.1 | NM_001940.3 | Refseq |

ATN1
Genomic Map