Assay Details
| Gene Symbol: MCHR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032503.2 | NP_115892.2 | 1 - 2 | 59 | 73 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001968.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_032503.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06024361.hg.1 | TC06001968.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06024361.hg.1 | ENST00000281806.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06024361.hg.1 | ENST00000445970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06024361.hg.1 | NM_001040179.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06024361.hg.1 | NM_032503.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06024356.hg.1 | TC06001968.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06024356.hg.1 | ENST00000281806.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06024356.hg.1 | ENST00000369212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06024356.hg.1 | ENST00000445970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06024356.hg.1 | NM_001040179.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06024356.hg.1 | NM_032503.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | ENST00000281806.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | ENST00000281806.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | ENST00000281806.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | ENST00000281806.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | ENST00000281806.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | ENST00000281806.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | ENST00000281806.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | ENST00000281806.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | ENST00000281806.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | ENST00000369212.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | ENST00000369212.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | ENST00000369212.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | ENST00000369212.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | ENST00000369212.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | ENST00000369212.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | ENST00000369212.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | NM_001040179.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | NM_001040179.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | NM_001040179.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | NM_001040179.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | NM_001040179.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | NM_001040179.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | NM_001040179.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | NM_001040179.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | NM_001040179.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | NM_032503.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | NM_032503.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | NM_032503.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | NM_032503.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | NM_032503.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | NM_032503.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | NM_032503.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012664.hg.1 | NM_032503.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600187349.hg.1 | ENST00000281806.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187349.hg.1 | ENST00000369212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187349.hg.1 | NM_001040179.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600187349.hg.1 | NM_032503.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600187349.hg.1 | ENST00000281806.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187349.hg.1 | ENST00000369212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187349.hg.1 | NM_001040179.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600187349.hg.1 | NM_032503.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600187349.hg.1 | TC0600012664.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600187353.hg.1 | ENST00000281806.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187353.hg.1 | ENST00000369212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187353.hg.1 | NM_001040179.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600187353.hg.1 | NM_032503.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600187353.hg.1 | ENST00000281806.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187353.hg.1 | ENST00000369212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600187353.hg.1 | NM_001040179.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600187353.hg.1 | NM_032503.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600187353.hg.1 | TC0600012664.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | ENST00000281806.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | ENST00000281806.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | ENST00000281806.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | ENST00000281806.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | ENST00000281806.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | ENST00000281806.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | ENST00000281806.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | ENST00000281806.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | ENST00000281806.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | ENST00000369212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | ENST00000369212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | ENST00000369212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | ENST00000369212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | ENST00000369212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | ENST00000369212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | ENST00000369212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | NM_001040179.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | NM_001040179.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | NM_001040179.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | NM_001040179.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | NM_001040179.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | NM_001040179.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | NM_001040179.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | NM_001040179.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | NM_001040179.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | NM_032503.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | NM_032503.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | NM_032503.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | NM_032503.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | NM_032503.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | NM_032503.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | NM_032503.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012664.hg.1 | NM_032503.2 | Refseq |

MCHR2
Genomic Map