Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012967.hg.1 |
ENST00000398955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012967.hg.1 |
ENST00000398955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012967.hg.1 |
ENST00000398955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012967.hg.1 |
ENST00000398955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012967.hg.1 |
ENST00000398955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012967.hg.1 |
ENST00000398955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012967.hg.1 |
ENST00000398955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012967.hg.1 |
ENST00000398955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012967.hg.1 |
NM_032623.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012967.hg.1 |
NM_032623.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012967.hg.1 |
NM_032623.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012967.hg.1 |
NM_032623.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012967.hg.1 |
NM_032623.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012967.hg.1 |
NM_032623.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012967.hg.1 |
NM_032623.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012967.hg.1 |
NM_032623.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022048.hg.1 |
TC04002948.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022048.hg.1 |
ENST00000398955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022048.hg.1 |
NM_032623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400191030.hg.1 |
ENST00000398955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400191030.hg.1 |
NM_032623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400191030.hg.1 |
ENST00000398955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400191030.hg.1 |
NM_032623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400191030.hg.1 |
TC0400012967.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400012967.hg.1 |
ENST00000398955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012967.hg.1 |
ENST00000398955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012967.hg.1 |
ENST00000398955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012967.hg.1 |
ENST00000398955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012967.hg.1 |
ENST00000398955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012967.hg.1 |
ENST00000398955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012967.hg.1 |
ENST00000398955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012967.hg.1 |
ENST00000398955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012967.hg.1 |
NM_032623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012967.hg.1 |
NM_032623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012967.hg.1 |
NM_032623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012967.hg.1 |
NM_032623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012967.hg.1 |
NM_032623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012967.hg.1 |
NM_032623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012967.hg.1 |
NM_032623.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012967.hg.1 |
NM_032623.3 |
Refseq |