Assay Details
| Gene Symbol: BBS9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014451.3 | NP_055266.2 | 13 - 14 | 1939 | 116 | |
| GenBank mRNA | AF095771.1 | - | 12 - 13 | 1420 | 116 | |
More Information
Set Membership: |
|

BBS9
Genomic Map