Assay Details
| Gene Symbol: CLEC1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099431.1 | NP_001092901.1 | 2 - 3 | 380 | 122 | |
| NM_016509.3 | NP_057593.3 | 3 - 4 | 479 | 122 | ||
| XM_005253382.4 | XP_005253439.1 | 5 - 6 | 851 | 122 | ||
| XM_011520685.2 | XP_011518987.1 | 4 - 5 | 760 | 122 | ||
| XM_011520686.2 | XP_011518988.1 | 3 - 4 | 388 | 122 | ||
| XM_017019395.1 | XP_016874884.1 | 4 - 5 | 835 | 122 | ||
| XM_017019396.1 | XP_016874885.1 | 2 - 3 | 187 | 122 | ||
| GenBank mRNA | AF124841.1 | - | 3 - 4 | 336 | 122 | |
| AY358599.1 | - | 3 - 4 | 460 | 122 | ||
| BC029554.1 | - | 4 - 5 | 451 | 5269252 | 122 | |
| BX647321.1 | - | 2 - 3 | 171 | 122 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001212.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12015962.hg.1 | ENST00000298527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12015962.hg.1 | ENST00000348658.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12015962.hg.1 | ENST00000398939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12015962.hg.1 | ENST00000428126.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12015962.hg.1 | NM_001099431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12015962.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12015962.hg.1 | TC12001212.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12015965.hg.1 | ENST00000298527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12015965.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12015965.hg.1 | TC12001212.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12015964.hg.1 | ENST00000298527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12015964.hg.1 | ENST00000348658.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12015964.hg.1 | ENST00000428126.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12015964.hg.1 | NM_001099431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12015964.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12015964.hg.1 | TC12001212.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12015966.hg.1 | ENST00000298527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12015966.hg.1 | ENST00000348658.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12015966.hg.1 | ENST00000428126.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12015966.hg.1 | NM_001099431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12015966.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12015966.hg.1 | TC12001212.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12015968.hg.1 | ENST00000298527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12015968.hg.1 | ENST00000348658.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12015968.hg.1 | ENST00000428126.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12015968.hg.1 | NM_001099431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12015968.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12015968.hg.1 | TC12001212.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | ENST00000298527.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | ENST00000298527.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | ENST00000298527.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | ENST00000298527.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | ENST00000298527.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | ENST00000298527.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | ENST00000298527.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | ENST00000298527.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | ENST00000348658.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | ENST00000348658.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | ENST00000348658.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | ENST00000348658.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | ENST00000348658.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | ENST00000348658.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | ENST00000348658.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | ENST00000398937.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | ENST00000398937.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | ENST00000398937.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | NM_001099431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | NM_001099431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | NM_001099431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | NM_001099431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | NM_001099431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | NM_001099431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | NM_001099431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009888.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174495.hg.1 | ENST00000298527.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174495.hg.1 | ENST00000348658.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174495.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174495.hg.1 | NM_001099431.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174495.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174495.hg.1 | ENST00000298527.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174495.hg.1 | ENST00000348658.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174495.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174495.hg.1 | NM_001099431.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174495.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174495.hg.1 | TC1200009888.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200174497.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174497.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174497.hg.1 | TC1200009888.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200174493.hg.1 | ENST00000298527.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174493.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174493.hg.1 | ENST00000298527.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174493.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174493.hg.1 | TC1200009888.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200174490.hg.1 | ENST00000298527.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174490.hg.1 | ENST00000348658.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174490.hg.1 | ENST00000398937.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174490.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174490.hg.1 | NM_001099431.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174490.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174490.hg.1 | ENST00000298527.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174490.hg.1 | ENST00000348658.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174490.hg.1 | ENST00000398937.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174490.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174490.hg.1 | NM_001099431.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174490.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174490.hg.1 | TC1200009888.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200174492.hg.1 | ENST00000298527.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174492.hg.1 | ENST00000348658.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174492.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174492.hg.1 | NM_001099431.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174492.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174492.hg.1 | ENST00000298527.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174492.hg.1 | ENST00000348658.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174492.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174492.hg.1 | NM_001099431.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174492.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174492.hg.1 | TC1200009888.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | ENST00000298527.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | ENST00000298527.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | ENST00000298527.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | ENST00000298527.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | ENST00000298527.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | ENST00000298527.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | ENST00000298527.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | ENST00000298527.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | ENST00000348658.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | ENST00000348658.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | ENST00000348658.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | ENST00000348658.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | ENST00000348658.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | ENST00000348658.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | ENST00000348658.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | ENST00000398937.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | ENST00000398937.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | ENST00000398937.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | NM_001099431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | NM_001099431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | NM_001099431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | NM_001099431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | NM_001099431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | NM_001099431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | NM_001099431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009888.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174494.hg.1 | ENST00000298527.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174494.hg.1 | ENST00000348658.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174494.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174494.hg.1 | NM_001099431.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174494.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174494.hg.1 | ENST00000298527.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174494.hg.1 | ENST00000348658.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174494.hg.1 | ENST00000428126.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174494.hg.1 | NM_001099431.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174494.hg.1 | NM_016509.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174494.hg.1 | TC1200009888.hg.1 | Clariom D TC |

CLEC1B
Genomic Map