Assay Details
| Gene Symbol: XPO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022459.4 | NP_071904.4 | 2 - 3 | 207 | 112 | |
| XM_005266503.3 | XP_005266560.1 | 2 - 3 | 207 | 112 | ||
| XM_011535198.1 | XP_011533500.1 | 2 - 3 | 188 | 112 | ||
| GenBank mRNA | AB051508.1 | - | 2 - 3 | 170 | 112 | |
| AK057924.1 | - | 2 - 3 | 178 | 112 | ||
| BC113571.1 | - | 1 - 2 | 110 | 8322623 | 112 | |
| BC113573.1 | - | 1 - 2 | 110 | 8322625 | 112 | |
| DA119417.1 | - | 2 - 3 | 207 | 112 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000461.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_011535198.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000465018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000465018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000465018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005272.hg.1 | ENST00000255305.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005272.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005272.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005272.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005272.hg.1 | TC13000461.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000465018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000465018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000465018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008267.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155826.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155826.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155826.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155826.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155826.hg.1 | ENST00000255305.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155826.hg.1 | ENST00000400602.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155826.hg.1 | ENST00000490513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155826.hg.1 | NM_022459.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155826.hg.1 | TC1300008267.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 167606 | - | - |

XPO4
Genomic Map