Assay Details
| Gene Symbol: MYH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002470.3 | NP_002461.2 | 33 - 34 | 4740 | 59 | |
| XM_011523870.2 | XP_011522172.1 | 33 - 34 | 6107 | 59 | ||
| XM_011523871.2 | XP_011522173.1 | 33 - 34 | 4740 | 59 | ||
| GenBank mRNA | AY517555.1 | - | 10 - 11 | 1656 | 59 | |
| CD614924.1 | - | 2 - 3 | 276 | 59 | ||
| X13100.1 | - | 13 - 14 | 2333 | 59 | ||
| X13988.1 | - | 32 - 33 | 4736 | 59 | ||
| X15696.1 | - | 12 - 13 | 2087 | 59 | ||
| X51593.1 | - | 12 - 13 | 2087 | 59 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001135.hg.1
Set Membership: |
|

MYH3
Genomic Map