Assay Details
| Gene Symbol: CNPPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321389.1 | NP_001308318.1 | 1 - 2 | 410 | 70 | |
| GenBank mRNA | AF151815.1 | - | 1 - 2 | 315 | 70 | |
| CX166359.1 | - | 1 - 2 | 406 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF151815.1, NM_001321389.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000409789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000409789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000409789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000409789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000409789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000409789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000409789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000409789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000409789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000409789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000409789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000409789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000409789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000451647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000451647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000451647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000451647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000451647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000451647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000451647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000451647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000451647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000451647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000453038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000453038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000453038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000453038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000453038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000453038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000453038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000453038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000453038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | ENST00000453038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000360507.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000409789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000409789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000409789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000409789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000409789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000409789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000409789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000409789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000409789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000409789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000409789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000409789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000409789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000451647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000451647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000451647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000451647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000451647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000451647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000451647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000451647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000451647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000451647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000453038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000453038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000453038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000453038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000453038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000453038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000453038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000453038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000453038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | ENST00000453038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015764.hg.1 | NM_015680.5 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 217057 | - | - |

CNPPD1
Genomic Map